siRNA Details
| Gene Symbol: R3hdm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134867.1 | NP_001128339.1 | 24 | 3290 | ||
| XM_006249690.2 | XP_006249752.1 | 24 | 3650 | |||
| XM_006249694.2 | XP_006249756.1 | 22 | 3155 | |||
| XM_006249698.1 | XP_006249760.1 | 17 | 2373 | |||
| XM_008769459.1 | XP_008767681.1 | 24 | 3239 | |||
| XM_008769460.1 | XP_008767682.1 | 23 | 3185 | |||
| XM_008769461.1 | XP_008767683.1 | 23 | 3032 | |||
| XM_008769462.1 | XP_008767684.1 | 23 | 2984 | |||
| XM_008769463.1 | XP_008767685.1 | 20 | 2797 | |||

R3hdm1
Genomic Map