siRNA Details
| Gene Symbol: Dyrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024767.1 | NP_001019938.1 | 3 | 1739 | ||
| XM_008769464.1 | XP_008767686.1 | 3 | 1951 | |||
| XM_008769465.1 | XP_008767687.1 | 2 | 1520 | |||
| GenBank mRNA | BC097474.1 | - | 3 | 1739 | 7457808 | |

Dyrk3
Genomic Map