siRNA Details
| Gene Symbol: Slc45a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135868.1 | NP_001129340.1 | 3, 4 | 1291 | ||
| XM_006249765.2 | XP_006249827.1 | 3, 4 | 1266 | |||
| XM_006249766.2 | XP_006249828.1 | 3, 4 | 1391 | |||
| XM_006249767.2 | XP_006249829.1 | 3, 4 | 1869 | |||
| XM_006249769.2 | XP_006249831.1 | 3, 4 | 2041 | |||

Slc45a3
Genomic Map