siRNA Details
| Gene Symbol: Mdm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012026.1 | NP_001012026.1 | 11 | 1275 | ||
| XM_006249777.2 | XP_006249839.1 | 11 | 1336 | |||
| XM_006249778.2 | XP_006249840.1 | 10 | 1193 | |||
| XM_006249779.2 | XP_006249841.1 | 10 | 1190 | |||
| GenBank mRNA | BC083647.1 | - | 11 | 1275 | 7135053 | |

Mdm4
Genomic Map