siRNA Details
| Gene Symbol: Elf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024768.1 | NP_001019939.1 | 6, 7 | 866 | ||
| XM_006249881.2 | XP_006249943.1 | 6, 7 | 961 | |||
| XM_006249883.2 | XP_006249945.1 | 6, 7 | 892 | |||
| GenBank mRNA | BC097980.1 | - | 6, 7 | 866 | 7443278 | |

Elf3
Genomic Map