siRNA Details
| Gene Symbol: Kcnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198762.1 | NP_942057.1 | 11 | 1067 | ||
| XM_006249942.2 | XP_006250004.1 | 11 | 1391 | |||
| XM_006249943.2 | XP_006250005.1 | 11 | 1391 | |||
| XM_008769564.1 | XP_008767786.1 | 12 | 1466 | |||
| GenBank mRNA | AY359443.1 | - | 11 | 1067 | ||

Kcnt2
Genomic Map