siRNA Details
| Gene Symbol: Trove2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107183.1 | NP_001100653.1 | 2 | 364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trove2| Gene Symbol: Trove2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107183.1 | NP_001100653.1 | 2 | 364 | ||