siRNA Details
| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013984.1 | NP_001014006.1 | 10, 11 | 814 | ||
| XM_006249997.2 | XP_006250059.1 | 9, 10 | 869 | |||
| GenBank mRNA | BC082004.1 | - | 10, 11 | 814 | 7131519 | |

Npl
Genomic Map