siRNA Details
| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013984.1 | NP_001014006.1 | 8 | 643 | ||
| XM_006249997.2 | XP_006250059.1 | 7 | 698 | |||
| GenBank mRNA | BC082004.1 | - | 8 | 643 | 7131519 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npl| Gene Symbol: Npl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013984.1 | NP_001014006.1 | 8 | 643 | ||
| XM_006249997.2 | XP_006250059.1 | 7 | 698 | |||
| GenBank mRNA | BC082004.1 | - | 8 | 643 | 7131519 | |