siRNA Details
| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 2 | 401 | ||
| XM_006249972.1 | 2 | 424 | ||||
| GenBank mRNA | BC101883.1 | - | 1 | 325 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpr| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 2 | 401 | ||
| XM_006249972.1 | 2 | 424 | ||||
| GenBank mRNA | BC101883.1 | - | 1 | 325 | ||