siRNA Details
Gene Symbol: Tpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107185.1 | NP_001100655.2 | 7 | 973 | ||
XM_006249972.1 | 7 | 996 | ||||
GenBank mRNA | BC101883.1 | - | 6 | 897 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107185.1 | NP_001100655.2 | 7 | 973 | ||
XM_006249972.1 | 7 | 996 | ||||
GenBank mRNA | BC101883.1 | - | 6 | 897 |