siRNA Details
| Gene Symbol: Tor1aip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199100.2 | NP_954531.1 | 5 | 1369 | ||
| XM_006250043.2 | XP_006250105.1 | 4 | 1274 | |||
| XM_006250044.2 | XP_006250106.1 | 4 | 1272 | |||
| XM_006250045.2 | XP_006250107.1 | 4 | 1272 | |||
| GenBank mRNA | BC061831.1 | - | 5 | 1326 | 5599615 | |

Tor1aip2
Genomic Map