siRNA Details
| Gene Symbol: Abl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107186.1 | NP_001100656.1 | 13 | 3437 | ||
| XM_006250046.2 | XP_006250108.1 | 12 | 3799 | |||
| XM_006250047.2 | XP_006250109.1 | 12 | 3796 | |||
| XM_006250048.2 | XP_006250110.1 | 12 | 4470 | |||
| XM_006250049.2 | XP_006250111.1 | 12 | 4466 | |||
| XM_006250050.2 | XP_006250112.1 | 11 | 3736 | |||
| XM_006250051.2 | XP_006250113.1 | 11 | 3733 | |||
| XM_006250052.2 | XP_006250114.1 | 13 | 3487 | |||
| XM_006250053.2 | XP_006250115.1 | 13 | 3483 | |||
| XM_006250054.2 | XP_006250116.1 | 13 | 4153 | |||
| XM_006250055.2 | XP_006250117.1 | 12 | 3420 | |||

Abl2
Genomic Map