siRNA Details
| Gene Symbol: Rasal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107188.1 | NP_001100658.1 | 10 | 1820 | ||
| XM_006250065.2 | XP_006250127.1 | 12 | 2832 | |||
| XM_006250066.2 | XP_006250128.1 | 12 | 3441 | |||
| XM_006250067.2 | XP_006250129.1 | 12 | 4300 | |||
| XM_008769635.1 | XP_008767857.1 | 12 | 2832 | |||
| XM_008769636.1 | XP_008767858.1 | 12 | 2811 | |||
| XM_008769637.1 | XP_008767859.1 | 12 | 2811 | |||
| XM_008769638.1 | XP_008767860.1 | 13 | 2485 | |||
| XM_008769639.1 | XP_008767861.1 | 13 | 2464 | |||

Rasal2
Genomic Map