siRNA Details
Gene Symbol: Astn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170603.1 | NP_001164074.1 | 9 | 1872 | ||
XM_008769640.1 | XP_008767862.1 | 6 | 1290 | |||
XM_008769641.1 | XP_008767863.1 | 6 | 1265 |
