siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 3 | 592 | ||
| XM_008769647.1 | XP_008767869.1 | 3 | 537 | |||
| GenBank mRNA | AY321346.1 | - | 451 | |||
| BC088467.1 | - | 3 | 592 | 7127992 | ||
| FQ209388.1 | - | 3 | 610 | |||
| FQ209496.1 | - | 3 | 553 | |||
| FQ209598.1 | - | 3 | 578 | |||
| FQ210275.1 | - | 3 | 575 | |||
| FQ210716.1 | - | 3 | 611 | |||
| FQ210762.1 | - | 3 | 610 | |||
| FQ211013.1 | - | 3 | 616 | |||
| FQ211135.1 | - | 3 | 610 | |||
| FQ218285.1 | - | 3 | 604 | |||
| FQ218964.1 | - | 3 | 575 | |||
| FQ219160.1 | - | 3 | 610 | |||
| FQ219233.1 | - | 3 | 575 | |||

Serpinc1
Genomic Map