siRNA Details
| Gene Symbol: Zbtb37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107191.1 | NP_001100661.1 | 4 | 1263 | ||
| XM_006250110.2 | XP_006250172.1 | 3 | 1118 | |||
| XM_006250111.2 | XP_006250173.1 | 3 | 1121 | |||
| XM_008769648.1 | XP_008767870.1 | 4 | 1311 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_173424, AK166242, AK148347, AK084871, AK078748, and others.

Zbtb37
Genomic Map