siRNA Details
Gene Symbol: Gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100563.1 | NP_001094033.1 | 5 | 802 | ||
XM_008769650.1 | XP_008767872.1 | 4 | 1378 | |||
GenBank mRNA | BC086370.1 | - | 3 | 281 | ||
BC160825.1 | - | 5 | 802 |
