siRNA Details
| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047878.1 | NP_001041343.1 | 12 | 3363 | ||
| GenBank mRNA | AY321333.1 | - | 12 | 3363 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F5| Gene Symbol: F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047878.1 | NP_001041343.1 | 12 | 3363 | ||
| GenBank mRNA | AY321333.1 | - | 12 | 3363 | ||