siRNA Details
| Gene Symbol: Pbx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100681.1 | NP_001094151.1 | 5, 6 | 1094 | ||
| NM_001134862.1 | NP_001128334.1 | 5, 6 | 1264 | |||
| GenBank mRNA | BC161983.1 | - | 5, 6 | 1094 | 9030389 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_183355, NM_008783, BC058390, AB221563, AK087587, and others.

Pbx1
Genomic Map