siRNA Details
| Gene Symbol: Igsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014787.1 | NP_001014787.1 | 4 | 923 | ||
| XM_006250286.2 | XP_006250348.1 | 4 | 1082 | |||
| XM_006250288.2 | XP_006250350.1 | 3 | 1024 | |||
| XM_006250289.2 | XP_006250351.1 | 3 | 861 | |||
| GenBank mRNA | BC092192.1 | - | 4 | 923 | 7312924 | |

Igsf8
Genomic Map