siRNA Details
| Gene Symbol: Tagln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013127.1 | NP_001013145.1 | 3 | 251 | ||
| XM_006250295.2 | XP_006250357.1 | 3 | 686 | |||
| GenBank mRNA | BC084703.1 | - | 3 | 251 | 7300611 | |
| FQ221577.1 | - | 3 | 262 | |||
| FQ223112.1 | - | 3 | 276 | |||
| FQ228757.1 | - | 3 | 270 | |||

Tagln2
Genomic Map