siRNA Details
| Gene Symbol: Aim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250335.1 | XP_006250397.1 | 4 | 953 | ||
| XM_006250336.2 | XP_006250398.1 | 5 | 1397 | |||
| XM_006250339.1 | XP_006250401.1 | 3 | 768 | |||
| XM_008769802.1 | XP_008768024.1 | 5 | 2612 | |||
| XM_008769803.1 | XP_008768025.1 | 5 | 3562 | |||
| XM_008769804.1 | XP_008768026.1 | 6 | 2940 | |||
| XM_008769805.1 | XP_008768027.1 | 5 | 2549 | |||
| XM_222949.8 | XP_222949.5 | 4 | 3377 | |||

Aim2
Genomic Map