siRNA Details
| Gene Symbol: Sccpdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013985.1 | NP_001014007.1 | 6 | 761 | ||
| GenBank mRNA | BC061579.1 | - | 565 | |||
| BC079215.1 | - | 6 | 761 | 7131885 | ||
| Gene Symbol: LOC103689999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769879.1 | XP_008768101.1 | 6 | 943 | ||

Sccpdh
Genomic Map