siRNA Details
| Gene Symbol: Prox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107201.1 | NP_001100671.1 | 2 | 1451 | ||
| XM_006250454.2 | XP_006250516.1 | 2 | 1577 | |||
| XM_006250455.2 | XP_006250517.1 | 2 | 1274 | |||
| XM_006250456.2 | XP_006250518.1 | 2 | 1896 | |||
| XM_006250458.2 | XP_006250520.1 | 2 | 1577 | |||

Prox1
Genomic Map