siRNA Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250494.2 | XP_006250556.1 | 6 | 515 | ||
| XM_223074.7 | XP_223074.5 | 6 | 674 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtl| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250494.2 | XP_006250556.1 | 6 | 515 | ||
| XM_223074.7 | XP_223074.5 | 6 | 674 | |||