siRNA Details
| Gene Symbol: Diexf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013986.2 | NP_001014008.1 | 4 | 512 | ||
| XM_006250482.2 | XP_006250544.1 | 5 | 702 | |||
| GenBank mRNA | BC087114.1 | - | 4 | 466 | 7193786 | |
| CB569988.1 | - | 524 | ||||
| FQ217506.1 | - | 5 | 685 | |||

Diexf
Genomic Map