siRNA Details
| Gene Symbol: GAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022041.3 | NP_071324.1 | 9, 10 | 1661 | ||
| XM_017023734.1 | XP_016879223.1 | 8, 9 | 1571 | |||
| GenBank mRNA | AF291673.1 | - | 9, 10 | 1657 | ||
| AK095378.1 | - | 9, 10 | 1661 | |||
| BC044840.1 | - | 9, 10 | 1648 | 2900666 | ||
| KU178409.1 | - | 9, 10 | 1513 | |||
| KU178410.1 | - | 7, 8 | 1047 | |||

GAN
Genomic Map