siRNA Details
| Gene Symbol: Zfp281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012030.1 | NP_001012030.1 | 2555 | |||
| GenBank mRNA | BC083844.1 | - | 2555 | 7121097 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp281| Gene Symbol: Zfp281 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012030.1 | NP_001012030.1 | 2555 | |||
| GenBank mRNA | BC083844.1 | - | 2555 | 7121097 | ||