siRNA Details
| Gene Symbol: Col9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100842.1 | NP_001094312.1 | 35 | 2550 | ||
| GenBank mRNA | DY313373.1 | - | 2 | 147 | ||
| DY316977.1 | - | 9 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col9a1| Gene Symbol: Col9a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100842.1 | NP_001094312.1 | 35 | 2550 | ||
| GenBank mRNA | DY313373.1 | - | 2 | 147 | ||
| DY316977.1 | - | 9 | 527 | |||