siRNA Details
| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107203.3 | NP_001100673.1 | 3 | 468 | ||
| GenBank mRNA | BC167003.1 | - | 3 | 469 | 8366732 | |
| FQ223120.1 | - | 2 | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo28| Gene Symbol: Fbxo28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107203.3 | NP_001100673.1 | 3 | 468 | ||
| GenBank mRNA | BC167003.1 | - | 3 | 469 | 8366732 | |
| FQ223120.1 | - | 2 | 164 | |||