siRNA Details
| Gene Symbol: LOC100364240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250596.2 | XP_006250658.2 | 9, 10 | 1295 | ||
| XM_006250597.2 | XP_006250659.2 | 9, 10 | 1295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100364240| Gene Symbol: LOC100364240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006250596.2 | XP_006250658.2 | 9, 10 | 1295 | ||
| XM_006250597.2 | XP_006250659.2 | 9, 10 | 1295 | |||