siRNA Details
| Gene Symbol: Zfp644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139484.1 | NP_001132956.1 | 7 | 3099 | ||
| XM_006250550.1 | XP_006250612.1 | 2 | 2786 | |||
| XM_006250554.2 | XP_006250616.1 | 2 | 2796 | |||
| XM_008769931.1 | XP_008768153.1 | 2 | 2860 | |||
| XM_008769932.1 | XP_008768154.1 | 3 | 2690 | |||
| XM_008769933.1 | XP_008768155.1 | 2 | 2786 | |||
| XM_008769934.1 | XP_008768156.1 | 3 | 2764 | |||
| XM_008769935.1 | XP_008768157.1 | 2 | 2786 | |||

Zfp644
Genomic Map