siRNA Details
| Gene Symbol: Lrrc8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107204.2 | NP_001100674.1 | 1 | 1485 | ||
| XM_006250557.1 | XP_006250619.1 | 4 | 1796 | |||
| GenBank mRNA | FQ212117.1 | - | 4 | 1801 | ||
| FQ217980.1 | - | 4 | 1751 | |||

Lrrc8b
Genomic Map