siRNA Details
| Gene Symbol: Aff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107206.1 | NP_001100676.1 | 3 | 344 | ||
| XM_006250632.2 | XP_006250694.1 | 4 | 449 | |||
| XM_006250633.2 | XP_006250695.1 | 3 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aff1| Gene Symbol: Aff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107206.1 | NP_001100676.1 | 3 | 344 | ||
| XM_006250632.2 | XP_006250694.1 | 4 | 449 | |||
| XM_006250633.2 | XP_006250695.1 | 3 | 333 | |||