siRNA Details
| Gene Symbol: Agpat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025670.1 | NP_001020841.1 | 11 | 1476 | ||
| XM_008770019.1 | XP_008768241.1 | 12 | 1473 | |||
| XM_008770020.1 | XP_008768242.1 | 12 | 1902 | |||
| XM_008770021.1 | XP_008768243.1 | 12 | 1601 | |||
| GenBank mRNA | BC097362.1 | - | 11 | 1476 | 7442656 | |

Agpat9
Genomic Map