siRNA Details
| Gene Symbol: Stbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013988.1 | NP_001014010.1 | 2 | 976 | ||
| GenBank mRNA | BC089867.1 | - | 2 | 976 | 7321620 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stbd1| Gene Symbol: Stbd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013988.1 | NP_001014010.1 | 2 | 976 | ||
| GenBank mRNA | BC089867.1 | - | 2 | 976 | 7321620 | |