siRNA Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012035.1 | NP_001012035.1 | 3 | 791 | ||
| XM_006250737.2 | XP_006250799.1 | 3 | 919 | |||
| XM_006250738.2 | XP_006250800.1 | 4 | 1028 | |||
| XM_006250739.2 | XP_006250801.1 | 3 | 1419 | |||
| XM_006250740.2 | XP_006250802.1 | 4 | 1480 | |||
| XM_006250741.2 | XP_006250803.1 | 3 | 1371 | |||
| XM_008770028.1 | XP_008768250.1 | 4 | 1528 | |||
| XM_008770029.1 | XP_008768251.1 | 3 | 1375 | |||
| GenBank mRNA | AY383681.1 | - | 2 | 292 | ||
| BC083590.1 | - | 3 | 791 | 7132550 | ||

Cdkl2
Genomic Map