siRNA Details
| Gene Symbol: Uba6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107213.1 | NP_001100683.1 | 2 | 177 | ||
| GenBank mRNA | FQ212356.1 | - | 2 | 162 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uba6| Gene Symbol: Uba6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107213.1 | NP_001100683.1 | 2 | 177 | ||
| GenBank mRNA | FQ212356.1 | - | 2 | 162 | ||