siRNA Details
Gene Symbol: Btf3l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238579.2 | XP_006238641.1 | 4, 5 | 485 | ||
XM_008763951.1 | XP_008762173.1 | 3, 4 | 429 | |||
XM_345561.7 | XP_345562.4 | 4, 5 | 496 | |||
GenBank mRNA | FQ222511.1 | - | 3, 4 | 367 | ||
FQ222777.1 | - | 4, 5 | 493 |
