siRNA Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107214.1 | NP_001100684.1 | 5 | 1317 | ||
| XM_006250898.2 | XP_006250960.1 | 5 | 1389 | |||
| XM_006250899.2 | XP_006250961.1 | 5 | 1388 | |||
| GenBank mRNA | BC158815.1 | - | 3 | 390 | ||

Slain2
Genomic Map