siRNA Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770169.1 | XP_008768391.1 | 4 | 409 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Apbb2| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770169.1 | XP_008768391.1 | 4 | 409 | ||