siRNA Details
| Gene Symbol: Rbm47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005882.1 | NP_001005882.1 | 2, 3 | 1278 | ||
| XM_006250949.2 | XP_006251011.1 | 3, 4 | 2361 | |||
| XM_006250950.2 | XP_006251012.1 | 3, 4 | 3259 | |||
| XM_006250951.2 | XP_006251013.1 | 5, 6 | 1607 | |||
| XM_006250952.2 | XP_006251014.1 | 4, 5 | 1833 | |||
| XM_006250954.2 | XP_006251016.1 | 4, 5 | 2364 | |||
| XM_006250955.2 | XP_006251017.1 | 3, 4 | 2552 | |||
| XM_008770152.1 | XP_008768374.1 | 3, 4 | 2066 | |||
| GenBank mRNA | AY724525.1 | - | 3, 4 | 1301 | ||
| BC081995.1 | - | 2, 3 | 1278 | 7130530 | ||

Rbm47
Genomic Map