siRNA Details
| Gene Symbol: Rhoh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013430.1 | NP_001013448.1 | 3 | 841 | ||
| GenBank mRNA | BC091269.1 | - | 3 | 841 | 7381512 | |
| FQ225173.1 | - | 3 | 853 | |||
| FQ228192.1 | - | 3 | 929 | |||
| FQ232983.1 | - | 3 | 853 | |||

Rhoh
Genomic Map