siRNA Details
| Gene Symbol: Klhl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047093.1 | NP_001040558.1 | 7 | 1556 | ||
| XM_006250959.2 | XP_006251021.1 | 7 | 1212 | |||
| XM_008770157.1 | XP_008768379.1 | 7 | 1034 | |||
| GenBank mRNA | AY641809.1 | - | 7 | 1556 | ||

Klhl5
Genomic Map