siRNA Details
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107216.1 | NP_001100686.1 | 2 | 902 | ||
XM_008770184.1 | XP_008768406.1 | 2 | 913 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107216.1 | NP_001100686.1 | 2 | 902 | ||
XM_008770184.1 | XP_008768406.1 | 2 | 913 |