siRNA Details
| Gene Symbol: Ncapg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770223.1 | XP_008768445.1 | 3, 4 | 699 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ncapg| Gene Symbol: Ncapg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770223.1 | XP_008768445.1 | 3, 4 | 699 | ||