siRNA Details
| Gene Symbol: Lgi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107219.2 | NP_001100689.2 | 8 | 1104 | ||
| GenBank mRNA | BC168885.1 | - | 8 | 1104 | 7121391 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgi2| Gene Symbol: Lgi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107219.2 | NP_001100689.2 | 8 | 1104 | ||
| GenBank mRNA | BC168885.1 | - | 8 | 1104 | 7121391 | |