siRNA Details
| Gene Symbol: Anapc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107220.1 | NP_001100690.1 | 25 | 2031 | ||
| GenBank mRNA | BC086546.1 | - | 15 | 945 | ||
| FQ227240.1 | - | 7 | 463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Anapc4| Gene Symbol: Anapc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107220.1 | NP_001100690.1 | 25 | 2031 | ||
| GenBank mRNA | BC086546.1 | - | 15 | 945 | ||
| FQ227240.1 | - | 7 | 463 | |||