siRNA Details
| Gene Symbol: C1qtnf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107221.1 | NP_001100691.1 | 3 | 566 | ||
| XM_006251073.2 | XP_006251135.1 | 3 | 1124 | |||
| XM_006251075.2 | XP_006251137.1 | 3 | 1827 | |||
| XM_008770208.1 | XP_008768430.1 | 2 | 1064 | |||
| GenBank mRNA | BC166720.1 | - | 3 | 954 | 7932439 | |
| FQ220553.1 | - | 3 | 564 | |||

C1qtnf7
Genomic Map